Please login to your online account to display your discounted pricing

Mouse anti-S100A1, Clone: SPM354, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476902

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6271
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen S100A1
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P23297
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified bovine brain S100 protein
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Protein A purified
Clone SPM354
Concentration 1mg/mL
Conjugate Unlabeled