Please login to your online account to display your discounted pricing

Mouse anti-S100A1, Clone: SPM354, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476901

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Immunogen Purified bovine brain S100 protein
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM354
Concentration 1mg/mL
Gene Accession No. P23297
Antigen S100A1
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Isotype IgG2a, κ
Gene ID (Entrez) 6271
Format Purified
Purification Method Purified
Reconstitution Protein A purified
Primary or Secondary Primary