Please login to your online account to display your discounted pricing

Mouse anti-ROR gamma/RORC/NR1F3, Clone: 4F3-3C8-2B7, Novus Biologicals™

$300.00 - $483.00

Specifications

Antigen ROR gamma/RORC/NR1F3
Clone 4F3-3C8-2B7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was RORg.
Gene ID (Entrez) 6097
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14325001

 
novus biologicals canada
NBP14325001MG
0.1mg Each for $483.00

NBP14325003

 
novus biologicals canada
NBP1432500025MG
0.025mg Each for $300.00
Description & Specifications

Specifications

Antigen ROR gamma/RORC/NR1F3
Clone 4F3-3C8-2B7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was RORg.
Gene ID (Entrez) 6097
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Western Blotting