Please login to your online account to display your discounted pricing

Mouse anti-ROR gamma/RORC/NR1F3, Clone: 4F3-3C8-2B7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1432500025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14325003

  • / Each
 

Description & Specifications

Specifications

Quantity 0.025mg
Primary or Secondary Primary
Antigen ROR gamma/RORC/NR1F3
Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Mouse
Applications Western Blotting
Immunogen The immunogen for this antibody was RORg.
Isotype IgG1
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6097
Clone 4F3-3C8-2B7
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human