Please login to your online account to display your discounted pricing

Mouse anti-ROR gamma/RORC/NR1F3, Clone: 4F3-3C8-2B7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14325001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14325001

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Immunogen The immunogen for this antibody was RORg.
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone 4F3-3C8-2B7
Conjugate Unlabeled
Antigen ROR gamma/RORC/NR1F3
Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Mouse
Isotype IgG1
Purification Method Affinity Purified
Primary or Secondary Primary
Gene ID (Entrez) 6097
Concentration 0.5mg/mL
Cross Reactivity Human