Please login to your online account to display your discounted pricing

Mouse anti-Rituximab, Clone: 109H9.06, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX5010P100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX5010P05

  • / Each
 

Description & Specifications

Specifications

Antigen Rituximab
Format Purified
Formulation Tris-NaCl pH 8
Host Species Mouse
Applications ELISA
Immunogen Anti-Mabthera(Rituximab)
Isotype IgG1
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Clone 109H9.06
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Purification Method Ion Exchange Chromatography
Monoclonal or Polyclonal Monoclonal