missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Rituximab, Clone: 108F11.02, Novus Biologicals™

$309.05 - $332.66

Specifications

Antigen Rituximab
Clone 108F11.02
Concentration 0.5mg/mL
Immunogen Anti-Mabthera(Rituximab)
Formulation Tris-NaCl pH 8
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
DDX5011B05 Novus Biologicals Canada
DDX5011B100005MG
0.05mg Each for $332.66
DDX5011P05 Novus Biologicals Canada
DDX5011P100005MG
0.05mg Each for $309.05
Specifications Description & Specifications

Specifications

Rituximab
0.5mg/mL
Tris-NaCl pH 8
Primary
Murine
Unlabeled
Ion Exchange Chromatography
108F11.02
Anti-Mabthera(Rituximab)
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
Western Blot