Please login to your online account to display your discounted pricing

Mouse anti-Rituximab, Clone: 108F11.02, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX5011P100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX5011P05

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Formulation Tris-NaCl pH 8
Cross Reactivity Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Quantity 0.05mg
Immunogen Anti-Mabthera(Rituximab)
Regulatory Status RUO
Host Species Mouse
Antigen Rituximab
Purification Method Ion Exchange Chromatography
Concentration 0.5mg/mL
Isotype IgG1
Format Purified
Clone 108F11.02
Applications ELISA