Please login to your online account to display your discounted pricing

Mouse anti-Rituximab, Clone: 108F11.02, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX5011B100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX5011B05

  • / Each
 

Description & Specifications

Specifications

Antigen Rituximab
Applications Western Blot
Clone 108F11.02
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Purified
Formulation Tris-NaCl pH 8
Host Species Murine
Immunogen Anti-Mabthera(Rituximab)
Isotype IgG1
Purification Method Ion Exchange Chromatography
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human