Please login to your online account to display your discounted pricing

Mouse anti-Retinol Binding Protein RBP, Clone: G4E4, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Retinol Binding Protein RBP
Clone G4E4
Concentration 0.2mg/mL
Immunogen Retinol binding protein purified from human plasma
Gene Accession No. P02753
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23423101

 
novus biologicals canada
NBP23423101ML
0.1mL Each for $139.00

NBP23423105

 
novus biologicals canada
NBP23423105ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Retinol Binding Protein RBP
Clone G4E4
Concentration 0.2mg/mL
Immunogen Retinol binding protein purified from human plasma
Gene Accession No. P02753
Gene ID (Entrez) 5947
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting