Please login to your online account to display your discounted pricing

Mouse anti-Retinol Binding Protein RBP, Clone: G4E4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423105

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Retinol Binding Protein RBP
Gene Accession No. P02753
Host Species Mouse
Immunogen Retinol binding protein purified from human plasma
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5947
Clone G4E4
Conjugate Unlabeled
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Isotype IgG1, κ
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Protein A purified
Concentration 0.2mg/mL