Please login to your online account to display your discounted pricing

Mouse anti-Retinol Binding Protein RBP, Clone: G4E4, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462201

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Immunogen Retinol binding protein purified from human plasma
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Conjugate Unlabeled
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Gene Accession No. P02753
Format Purified
Purification Method Purified
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Clone G4E4
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Antigen Retinol Binding Protein RBP
Reconstitution Protein A purified
Regulatory Status RUO
Gene ID (Entrez) 5947