Please login to your online account to display your discounted pricing

Mouse anti-Renal Cell Carcinoma (gp200), Clone: MG38, Biotin, Novus Biologicals™

$192.00 - $354.00

Specifications

Antigen Renal Cell Carcinoma (gp200)
Clone MG38
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD205.
Gene ID (Entrez) 4065
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14356801

 
novus biologicals canada
NBP14356801MG
0.1mg Each for $354.00

NBP14356803

 
novus biologicals canada
NBP1435680025MG
0.025mg Each for $192.00
Description & Specifications

Specifications

Antigen Renal Cell Carcinoma (gp200)
Clone MG38
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD205.
Gene ID (Entrez) 4065
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry