Please login to your online account to display your discounted pricing

Mouse anti-RBP4/Retinol-Binding Protein 4, Clone: SPM442, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Clone SPM442
Concentration 0.2mg/mL
Immunogen Retinol binding protein purified from human plasma
Gene ID (Entrez) 5950
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23436201

 
novus biologicals canada
NBP23436201ML
0.1mL Each for $139.00

NBP23436205

 
novus biologicals canada
NBP23436205ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Clone SPM442
Concentration 0.2mg/mL
Immunogen Retinol binding protein purified from human plasma
Gene ID (Entrez) 5950
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting