Please login to your online account to display your discounted pricing

Mouse anti-RBP4/Retinol-Binding Protein 4, Clone: SPM442, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23436205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23436205

  • / Each
 

Description & Specifications

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM442
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Retinol binding protein purified from human plasma
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5950
Test Specificity Recognizes a protein of 21kDa-25kDa, identified as retinol binding protein (RBP). Its epitope localizes between aa 74-182 of human RBP. This MAb recognizes reduced and carboxy-methylated RBP (RCM-RBP) as well as the circulatory RBP but not the native RBP, thereby suggesting that its epitope becomes accessible either on unfolding or upon binding of RBP to transthyretin (prealbumin).
Reconstitution Protein A purified