Please login to your online account to display your discounted pricing

Mouse anti-RBP4/Retinol-Binding Protein 4, Clone: SPM442, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Clone SPM442
Concentration 1mg/mL
Immunogen Retinol binding protein purified from human plasma
Gene ID (Entrez) 5950
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23441001

 
novus biologicals canada
NBP23441001MG
0.1mg Each for $384.00

NBP23441002

 
novus biologicals canada
NBP23441002MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Clone SPM442
Concentration 1mg/mL
Immunogen Retinol binding protein purified from human plasma
Gene ID (Entrez) 5950
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Goat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting