Please login to your online account to display your discounted pricing

Mouse anti-Raf-1, Clone: 4G4, Novus Biologicals™

$367.12 - $397.00

Specifications

Antigen Raf-1
Clone 4G4
Concentration 1mg/mL
Immunogen Purified recombinant fragment of human RAF1 expressed in E. coli.
Gene ID (Entrez) 5894
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP237513

 
novus biologicals canada
NBP237513
0.1mL Each for $367.12

NBP237516

 
novus biologicals canada
NBP237516
0.1mL Each for $397.00
Description & Specifications

Specifications

Antigen Raf-1
Clone 4G4
Concentration 1mg/mL
Immunogen Purified recombinant fragment of human RAF1 expressed in E. coli.
Gene ID (Entrez) 5894
Formulation PBS
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Western Blotting