Please login to your online account to display your discounted pricing

Mouse anti-Raf-1, Clone: 4G4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP237516

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP237516

  • / Each
 

Description & Specifications

Specifications

Immunogen Purified recombinant fragment of human RAF1 expressed in E. coli.
Conjugate Unlabeled
Primary or Secondary Primary
Isotype IgG1
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Western Blotting
Cross Reactivity Human
Host Species Mouse
Quantity 0.1mL
Format Affinity Purified
Antigen Raf-1
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Formulation PBS
Concentration 1mg/mL
Gene ID (Entrez) 5894
Regulatory Status RUO
Clone 4G4