Please login to your online account to display your discounted pricing

Mouse anti-Qa-2, Clone: 69H1-9-9, Novus Biologicals™

$219.00 - $384.00

Specifications

Antigen Qa-2
Clone 69H1-9-9
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Qa2.
Gene ID (Entrez) 111458
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14325701

 
novus biologicals canada
NBP14325701MG
0.1mg Each for $274.00

NBP14325705

 
novus biologicals canada
NBP143257005MG
0.05mg Each for $219.00

NBP1432575

 
novus biologicals canada
NBP14325705MG
0.5mg Each for $384.00
Description & Specifications

Specifications

Antigen Qa-2
Clone 69H1-9-9
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Qa2.
Gene ID (Entrez) 111458
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay