Please login to your online account to display your discounted pricing

Mouse anti-Qa-2, Clone: 69H1-9-9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14325705MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432575

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 111458
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Quantity 0.5mg
Antigen Qa-2
Format Affinity Purified
Immunogen The immunogen for this antibody was Qa2.
Clone 69H1-9-9
Primary or Secondary Primary
Isotype IgG2a, κ
Conjugate Unlabeled
Cross Reactivity Mouse
Host Species Mouse
Regulatory Status RUO
Concentration 0.5mg/mL
Formulation Phosphate buffer pH 7.2