Please login to your online account to display your discounted pricing

Mouse anti-Qa-2, Clone: 69H1-9-9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14325701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14325701

  • / Each
 

Description & Specifications

Specifications

Immunogen The immunogen for this antibody was Qa2.
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Isotype IgG2a, κ
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 111458
Clone 69H1-9-9
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Antigen Qa-2
Purification Method Affinity Purified