Please login to your online account to display your discounted pricing

Mouse anti-Prolactin R, Clone: SPM213, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23289801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23289801

  • / Each
 

Description & Specifications

Specifications

Antigen Prolactin R
Gene Accession No. P16471
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Semi-purified human prolactin receptor
Isotype IgG1, κ
Format Purified
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 5618
Reconstitution Protein A purified
Clone SPM213
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Cross Reactivity Human