Please login to your online account to display your discounted pricing

Mouse anti-Prolactin R, Clone: SPM213, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474602

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Semi-purified human prolactin receptor
Gene Accession No. P16471
Test Specificity It recognizes a protein of 70kDa, identified as prolactin receptor.
Concentration 1mg/mL
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM213
Antigen Prolactin R
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Quantity 0.2mg
Conjugate Unlabeled
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1, κ
Gene ID (Entrez) 5618
Reconstitution Protein A purified
Format Purified
Monoclonal or Polyclonal Monoclonal