missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Prolactin R, Clone: PRLR742, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Prolactin R
Clone PRLR742
Concentration 1mg/mL
Immunogen Recombinant human prolactin receptor
Gene Accession No. P16471
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23471201 Novus Biologicals Canada
NBP23471201MG
0.1mg Each for $384.00
NBP23471202 Novus Biologicals Canada
NBP23471202MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Prolactin R
1mg/mL
P16471
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
PRLR742
Recombinant human prolactin receptor
5618
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot