Please login to your online account to display your discounted pricing

Mouse anti-Prolactin R, Clone: PRLR742, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471202

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen Prolactin R
Host Species Mouse
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 5618
Reconstitution Protein A purified
Clone PRLR742
Cross Reactivity Human
Gene Accession No. P16471
Immunogen Recombinant human prolactin receptor
Regulatory Status RUO
Concentration 1mg/mL