Please login to your online account to display your discounted pricing

Mouse anti-Prolactin R, Clone: PRLR742, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Prolactin R
Primary or Secondary Primary
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Immunogen Recombinant human prolactin receptor
Conjugate Unlabeled
Concentration 1mg/mL
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P16471
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 5618
Format Purified
Clone PRLR742
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Quantity 0.1mg