Please login to your online account to display your discounted pricing

Mouse anti-Prolactin R, Clone: B6.2+PRLR742, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Prolactin R
Clone B6.2+PRLR742
Concentration 0.2mg/mL
Immunogen Semi-purified human prolactin receptor (B6.2); Recombinant human prolactin receptor (PRLR742)
Gene Accession No. P16471
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23428601

 
novus biologicals canada
NBP23428601ML
0.1mL Each for $139.00

NBP23428605

 
novus biologicals canada
NBP23428605ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Prolactin R
Clone B6.2+PRLR742
Concentration 0.2mg/mL
Immunogen Semi-purified human prolactin receptor (B6.2); Recombinant human prolactin receptor (PRLR742)
Gene Accession No. P16471
Gene ID (Entrez) 5618
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting