Please login to your online account to display your discounted pricing

Mouse anti-Prolactin R, Clone: B6.2+PRLR742, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23428601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23428601

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Accession No. P16471
Reconstitution Protein A purified
Isotype IgG1, κ
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Concentration 0.2mg/mL
Monoclonal or Polyclonal Monoclonal
Immunogen Semi-purified human prolactin receptor (B6.2); Recombinant human prolactin receptor (PRLR742)
Clone B6.2+PRLR742
Gene ID (Entrez) 5618
Host Species Mouse
Antigen Prolactin R
Format Purified
Primary or Secondary Primary
Quantity 0.1mL