Please login to your online account to display your discounted pricing

Mouse anti-Prolactin R, Clone: B6.2+PRLR742, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23455501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23455501

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Prolactin R
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Semi-purified human prolactin receptor (B6.2); Recombinant human prolactin receptor (PRLR742)
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5618
Reconstitution Protein A purified
Clone B6.2+PRLR742
Cross Reactivity Human
Gene Accession No. P16471
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Conjugate Unlabeled