Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: SPM566, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Progesterone R/NR3C3
Clone SPM566
Concentration 0.2mg/mL
Immunogen Recombinant human Progesterone Receptor protein
Gene Accession No. P06401
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23438201

 
novus biologicals canada
NBP23438201ML
0.1mL Each for $139.00

NBP23438205

 
novus biologicals canada
NBP23438205ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Progesterone R/NR3C3
Clone SPM566
Concentration 0.2mg/mL
Immunogen Recombinant human Progesterone Receptor protein
Gene Accession No. P06401
Gene ID (Entrez) 5241
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting