Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: SPM566, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438201

  • / Each
 

Description & Specifications

Specifications

Antigen Progesterone R/NR3C3
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone SPM566
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P06401
Host Species Mouse
Immunogen Recombinant human Progesterone Receptor protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5241
Test Specificity This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family.
Reconstitution Protein A purified