Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: SPM566, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Progesterone R/NR3C3
Clone SPM566
Concentration 1mg/mL
Immunogen Recombinant human Progesterone Receptor protein
Gene Accession No. P06401
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23442901

 
novus biologicals canada
NBP23442901MG
0.1mg Each for $384.00

NBP23442902

 
novus biologicals canada
NBP23442902MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Progesterone R/NR3C3
Clone SPM566
Concentration 1mg/mL
Immunogen Recombinant human Progesterone Receptor protein
Gene Accession No. P06401
Gene ID (Entrez) 5241
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot