Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: SPM566, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442901

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P06401
Host Species Mouse
Immunogen Recombinant human Progesterone Receptor protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 5241
Reconstitution Protein A purified
Clone SPM566
Conjugate Unlabeled
Antigen Progesterone R/NR3C3
Monoclonal or Polyclonal Monoclonal
Test Specificity This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family.
Concentration 1mg/mL