missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: PR484, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP23332402MG

 View more versions of this product

Catalog No. NBP23332402


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Progesterone R/NR3C3
PR484
Unlabeled
PBS with 0.05% BSA. Protein G purified.
A progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped.
Purified
RUO
Primary
5241
Western Blot
0.2mg/mL
Purified
Murine
IgG1, κ
0.2mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human