Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: PR484, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23332502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23332502

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Quantity 0.2mg
Host Species Mouse
Primary or Secondary Primary
Format Purified
Regulatory Status RUO
Isotype IgG1, κ
Clone PR484
Concentration 1.0mg/mL
Monoclonal or Polyclonal Monoclonal
Formulation PBS. Does not contain BSA. Protein G purified.
Antigen Progesterone R/NR3C3
Storage Requirements Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Gene ID (Entrez) 5241
Immunogen A progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped.
Purification Method Purified