Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: PR484, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23332501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23332501

  • / Each
 

Description & Specifications

Specifications

Antigen Progesterone R/NR3C3
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone PR484
Concentration 1.0mg/mL
Conjugate Unlabeled
Format Purified
Formulation PBS. Does not contain BSA. Protein G purified.
Host Species Murine
Immunogen A progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5241
Target Species Human