Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: 1A6, Novus Biologicals™

$559.00 - $719.00

Specifications

Antigen Progesterone R/NR3C3
Clone 1A6
Immunogen Recombinant human progesterone protein.
Gene ID (Entrez) 5241
Formulation Tissue culture supernatant
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP22667403

 
novus biologicals canada
NBP226674025ML
0.25mL Each for $559.00

NBP22667405

 
novus biologicals canada
NBP22667405ML
0.5mL Each for $719.00
Description & Specifications

Specifications

Antigen Progesterone R/NR3C3
Clone 1A6
Immunogen Recombinant human progesterone protein.
Gene ID (Entrez) 5241
Formulation Tissue culture supernatant
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Supernatant
Purification Method Supernatant
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)