Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: 1A6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP22667405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP22667405

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Host Species Mouse
Quantity 0.5mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen Progesterone R/NR3C3
Formulation Tissue culture supernatant
Immunogen Recombinant human progesterone protein.
Isotype IgG1, κ
Purification Method Supernatant
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5241
Test Specificity This antibody is specific to human progesterone receptor protein. Progesterone receptor is an estrogen regulated protein and its expression indicates a responsive estrogen receptor pathway.
Clone 1A6
Format Supernatant