Please login to your online account to display your discounted pricing

Mouse anti-Progesterone R/NR3C3, Clone: 1A6, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP226674025ML

 View all options for this product


For Research Use Only

Catalog No. NBP22667403


Description & Specifications


Antigen Progesterone R/NR3C3
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone 1A6
Conjugate Unlabeled
Cross Reactivity Human
Format Supernatant
Formulation Tissue culture supernatant
Host Species Mouse
Immunogen Recombinant human progesterone protein.
Isotype IgG1, κ
Purification Method Supernatant
Quantity 0.25mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5241
Test Specificity This antibody is specific to human progesterone receptor protein. Progesterone receptor is an estrogen regulated protein and its expression indicates a responsive estrogen receptor pathway.