missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-PP14/Glycodelin, Clone: NA, Novus Biologicals™

$219.00 - $661.49

Specifications

Antigen PP14/Glycodelin
Clone NA
Concentration 1.0mg/mL
Immunogen Placental protein 14 (glycodelin A) purified from second trimester amniotic fluid
Gene ID (Entrez) 5047
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP10505112 Novus Biologicals Canada
NBP105051125MG
0.125mg Each for $219.00
NBP10505102 Novus Biologicals Canada
NBP105051025MG
0.25mg Each for $661.49
Specifications Description & Specifications

Specifications

PP14/Glycodelin
1.0mg/mL
5047
Monoclonal
Store at 4°C. Do not freeze.
IgG1, κ
Purified
ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
NA
Placental protein 14 (glycodelin A) purified from second trimester amniotic fluid
0.01M phosphate buffer, pH 7.4, with 0.5M NaCl
Primary
Murine
Unlabeled
Purified