Please login to your online account to display your discounted pricing

Mouse anti-PP14/Glycodelin, Clone: NA, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP105051025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP10505102

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Primary or Secondary Primary
Antigen PP14/Glycodelin
Format Purified
Formulation 0.01M phosphate buffer, pH 7.4, with 0.5M NaCl
Host Species Mouse
Immunogen Placental protein 14 (glycodelin A) purified from second trimester amniotic fluid
Isotype IgG1, κ
Purification Method Purified
Quantity 0.25mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5047
Test Specificity Placental protein 14 (human, glycodelin A)
Reconstitution Protein A purified
Clone NA
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human