Please login to your online account to display your discounted pricing

Mouse anti-PMEL17/SILV, Clone: SPM286, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen PMEL17/SILV
Clone SPM286
Concentration 0.2mg/mL
Immunogen Membranes from a human melanoma metastasis
Gene Accession No. P40967
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23285501

 
novus biologicals canada
NBP23285501ML
0.1mL Each for $139.00

NBP23285510

 
novus biologicals canada
NBP23285510ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen PMEL17/SILV
Clone SPM286
Concentration 0.2mg/mL
Immunogen Membranes from a human melanoma metastasis
Gene Accession No. P40967
Gene ID (Entrez) 6490
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Equine
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)