Please login to your online account to display your discounted pricing

Mouse anti-PMEL17/SILV, Clone: SPM286, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23285510ML

 View all options for this product


For Research Use Only

Catalog No. NBP23285510


Description & Specifications


Antigen PMEL17/SILV
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone SPM286
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Equine
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P40967
Host Species Mouse
Immunogen Membranes from a human melanoma metastasis
Isotype IgG2b, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6490
Test Specificity Does not react to Rat.
Reconstitution Protein A purified