missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-PMEL17/SILV, Clone: SPM286, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen PMEL17/SILV
Clone SPM286
Concentration 1mg/mL
Immunogen Membranes from a human melanoma metastasis
Gene Accession No. P40967
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23475502 Novus Biologicals Canada
NBP23475502MG
0.2mg Each for $527.00
NBP23475501 Novus Biologicals Canada
NBP23475501MG
0.1mg Each for $384.00
Specifications Description & Specifications

Specifications

PMEL17/SILV
1mg/mL
P40967
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM286
Membranes from a human melanoma metastasis
6490
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)