Please login to your online account to display your discounted pricing

Mouse anti-PMEL17/SILV, Clone: SPM286, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475501

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Monoclonal
Antigen PMEL17/SILV
Quantity 0.1mg
Cross Reactivity Equine
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Test Specificity This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Does not react with Rat.
Isotype IgG2b, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Format Purified
Primary or Secondary Primary
Gene Accession No. P40967
Host Species Mouse
Conjugate Unlabeled
Gene ID (Entrez) 6490
Immunogen Membranes from a human melanoma metastasis
Regulatory Status RUO
Clone SPM286
Concentration 1mg/mL
Purification Method Purified