Please login to your online account to display your discounted pricing

Mouse anti-PMEL17/SILV, Clone: SPM142, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23285401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23285401

  • / Each
 

Description & Specifications

Specifications

Antigen PMEL17/SILV
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Extract of pigmented melanoma metastases from lymph nodes
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P40967
Host Species Mouse
Isotype IgG1, κ
Quantity 0.1mL
Primary or Secondary Primary
Gene ID (Entrez) 6490
Test Specificity Does not react to Canine or Rat.
Reconstitution Protein A purified
Clone SPM142
Conjugate Unlabeled
Cross Reactivity Human
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL