Please login to your online account to display your discounted pricing

Mouse anti-PMEL17/SILV, Clone: SPM142, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23285410ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23285410

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen PMEL17/SILV
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P40967
Immunogen Extract of pigmented melanoma metastases from lymph nodes
Isotype IgG1, κ
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6490
Reconstitution Protein A purified
Concentration 0.2mg/mL
Host Species Mouse
Purification Method Purified
Test Specificity Does not react to Canine or Rat.
Clone SPM142
Conjugate Unlabeled