Please login to your online account to display your discounted pricing

Mouse anti-PMEL17/SILV, Clone: SPM142, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen PMEL17/SILV
Clone SPM142
Concentration 1mg/mL
Immunogen Extract of pigmented melanoma metastases from lymph nodes
Gene Accession No. P40967
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23473901

 
novus biologicals canada
NBP23473901MG
0.1mg Each for $384.00

NBP23473902

 
novus biologicals canada
NBP23473902MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen PMEL17/SILV
Clone SPM142
Concentration 1mg/mL
Immunogen Extract of pigmented melanoma metastases from lymph nodes
Gene Accession No. P40967
Gene ID (Entrez) 6490
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting