Please login to your online account to display your discounted pricing

Mouse anti-PMEL17/SILV, Clone: SPM142, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473902

  • / Each
 

Description & Specifications

Specifications

Antigen PMEL17/SILV
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM142
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P40967
Host Species Murine
Immunogen Extract of pigmented melanoma metastases from lymph nodes
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6490
Test Specificity This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Does not react with Canine or Rat.
Reconstitution Protein A purified
Target Species Human