Please login to your online account to display your discounted pricing

Mouse anti-PMEL17/SILV, Clone: HMB45, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463801

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Extract of pigmented melanoma metastases from lymph nodes
Purification Method Purified
Reconstitution Protein A purified
Cross Reactivity Human
Antigen PMEL17/SILV
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P40967
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6490
Test Specificity Does not react to Canine or Rat.
Clone HMB45
Concentration 1mg/mL
Conjugate Unlabeled